Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hey1
Entrez Gene ID
155437
Gene Name
hes-related family bHLH transcription factor with YRPW motif 1
Gene Aliases
Herp2
Chromosome Location
Chr.2: 95320283 - 95322701 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191845.1 NP_001178774.1 1 - 2 150 61

View Full Product Details