Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd31
Entrez Gene ID
100359720
Gene Name
ankyrin repeat domain 31
Gene Aliases
-
Chromosome Location
Chr.2: 27571133 - 27743163 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760712.1 XP_008758934.1 8 - 9 1428 124

View Full Product Details