The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035221.1 | NP_001030298.1 | - | 245 | 121 |
XM_008764508.1 | XP_008762730.1 | - | 452 | 121 | |
XM_008764509.1 | XP_008762731.1 | - | 700 | 121 | |
XM_008764510.1 | XP_008762732.1 | - | 651 | 121 | |
GenBank mRNA | CB584328.1 | - | - | 256 | 121 |
FQ229713.1 | - | - | 212 | 121 |