Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab25
Entrez Gene ID
310632
Gene Name
RAB25, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.2: 187903301 - 187909398 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107687.2 NP_001101157.2 4 - 5 737 64
GenBank mRNA BC168745.1 - 4 - 5 734 64

View Full Product Details