Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pou5f2
Entrez Gene ID
680620
Gene Name
POU domain class 5, transcription factor 2
Gene Aliases
Sprm-1, Sprm1
Chromosome Location
Chr.2: 5437067 - 5438301 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081751.1 NP_001075220.1 1 - 1 1053 105
GenBank mRNA L23864.1 - 1 - 1 1053 105

View Full Product Details