The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031097.1 | NP_112359.1 | 10 - 11 | 1799 | 129 |
GenBank mRNA | AY724503.1 | - | 10 - 11 | 1974 | 129 |
BC061718.1 | - | 10 - 11 | 1832 | 129 | |
D87515.1 | - | 10 - 11 | 1796 | 129 | |
U61696.1 | - | 10 - 11 | 1799 | 129 |