Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npl
Entrez Gene ID
304860
Gene Name
N-acetylneuraminate pyruvate lyase
Gene Aliases
-
Chromosome Location
Chr.13: 70938073 - 70981000 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013984.1 NP_001014006.1 3 - 4 213 95
XM_006249997.2 XP_006250059.1 3 - 4 361 95
GenBank mRNA BC082004.1 - 3 - 4 213 95

View Full Product Details