Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ercc1
Entrez Gene ID
292673
Gene Name
excision repair cross-complementation group 1
Gene Aliases
-
Chromosome Location
Chr.1: 80256973 - 80268198 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106228.1 NP_001099698.1 8 - 9 893 54
XM_006228395.2 XP_006228457.1 8 - 9 911 54
XM_006228396.1 XP_006228458.1 8 - 9 871 54
GenBank mRNA FQ225470.1 - 8 - 9 882 54

View Full Product Details