Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Znhit1
Entrez Gene ID
100360077
Gene Name
zinc finger, HIT-type containing 1
Gene Aliases
-
Chromosome Location
Chr.12: 22726643 - 22732917 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006249091.2 XP_006249153.1 1 - 2 127 121

View Full Product Details