The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134766.1 | NP_001128238.1 | 9 - 10 | 1831 | 98 |
XM_006249355.2 | XP_006249417.1 | 9 - 10 | 2699 | 98 | |
XM_006249356.2 | XP_006249418.1 | 9 - 10 | 2645 | 98 | |
XM_006249357.2 | XP_006249419.1 | 9 - 10 | 2699 | 98 | |
XM_006249358.1 | XP_006249420.1 | 9 - 10 | 1772 | 98 | |
XM_008769265.1 | XP_008767487.1 | 8 - 9 | 1584 | 98 | |
GenBank mRNA | BC086549.1 | - | 5 - 6 | 1008 | 98 |
BC099139.1 | - | 5 - 6 | 1110 | 98 |