Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mospd3
Entrez Gene ID
288557
Gene Name
motile sperm domain containing 3
Gene Aliases
-
Chromosome Location
Chr.12: 22164983 - 22169251 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025629.1 NP_001020800.1 1 - 2 327 79
XM_006249115.2 XP_006249177.1 2 - 3 387 79
GenBank mRNA BC099230.1 - 1 - 2 327 79

View Full Product Details