Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oas2
Entrez Gene ID
363938
Gene Name
2'-5' oligoadenylate synthetase 2
Gene Aliases
-
Chromosome Location
Chr.12: 41341417 - 41363124 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009715.1 NP_001009715.1 9 - 10 2026 66
GenBank mRNA AY230746.1 - 9 - 10 2026 66

View Full Product Details