The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009379.1 | NP_001009379.1 | - | 1451 | 113 |
GenBank mRNA | AY196699.1 | - | - | 1451 | 113 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008769299.1 | XP_008767521.1 | 6 - 6 | 1551 | 113 |