Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsga13
Entrez Gene ID
688848
Gene Name
testis specific, 13
Gene Aliases
-
Chromosome Location
Chr.4: 58195424 - 58216712 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107851.1 NP_001101321.1 3 - 4 322 132

View Full Product Details