Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbm4
Entrez Gene ID
293663
Gene Name
RNA binding motif protein 4
Gene Aliases
-
Chromosome Location
Chr.1: 220038403 - 220049748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170484.1 NP_001163955.1 3 - 4 1204 57
XM_008760101.1 XP_008758323.1 3 - 4 2128 57
GenBank mRNA FQ212306.1 - 3 - 4 1310 57

View Full Product Details