Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scgb1d2
Entrez Gene ID
309203
Gene Name
secretoglobin, family 1D, member 2
Gene Aliases
Pbpc1bs, Psbpc1
Chromosome Location
Chr.1: 225591848 - 225600915 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177482.1 NP_803435.1 2 - 3 267 80
XM_008760227.1 XP_008758449.1 4 - 5 591 80
XM_008760228.1 XP_008758450.1 4 - 5 722 80
GenBank mRNA BC099696.1 - 2 - 3 247 80
V01545.1 - 2 - 3 267 80

View Full Product Details