The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012170.1 | NP_001012170.1 | 14 - 15 | 1235 | 62 |
XM_006230823.2 | XP_006230885.1 | 14 - 15 | 1241 | 62 | |
XM_006230824.2 | XP_006230886.1 | 15 - 16 | 1308 | 62 | |
GenBank mRNA | BC085801.1 | - | 14 - 15 | 1235 | 62 |