Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppm1h
Entrez Gene ID
314897
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1H
Gene Aliases
-
Chromosome Location
Chr.7: 66845692 - 67117317 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC088307.1 - 4 - 5 531 127

View Full Product Details