Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gsdmd
Entrez Gene ID
315084
Gene Name
gasdermin D
Gene Aliases
Gsdmdc1
Chromosome Location
Chr.7: 116889152 - 116894140 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130553.1 NP_001124025.1 5 - 6 772 74
XM_006241809.2 XP_006241871.1 5 - 6 1427 74

View Full Product Details