Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdyhv1
Entrez Gene ID
362914
Gene Name
WDYHV motif containing 1
Gene Aliases
RGD1311362
Chromosome Location
Chr.7: 97957022 - 97977135 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025024.1 NP_001020195.1 1 - 2 229 98
GenBank mRNA BC091306.1 - 1 - 2 229 98

View Full Product Details