Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nipal2
Entrez Gene ID
362899
Gene Name
NIPA-like domain containing 2
Gene Aliases
Npal2, RGD1564315
Chromosome Location
Chr.7: 73339706 - 73450262 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130559.1 NP_001124031.1 6 - 7 1155 157

View Full Product Details