Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpys
Entrez Gene ID
65135
Gene Name
dihydropyrimidinase
Gene Aliases
-
Chromosome Location
Chr.7: 78769918 - 78847941 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC081768.1 - 9 - 10 1639 163

View Full Product Details