Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam91a1
Entrez Gene ID
689997
Gene Name
family with sequence similarity 91, member A1
Gene Aliases
-
Chromosome Location
Chr.7: 98302802 - 98341201 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127578.1 NP_001121050.1 3 - 4 524 108
XM_006241684.2 XP_006241746.1 3 - 4 528 108
GenBank mRNA BC166852.1 - 3 - 4 524 108

View Full Product Details