Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olr53
Entrez Gene ID
405378
Gene Name
olfactory receptor 53
Gene Aliases
-
Chromosome Location
Chr.1: 167970207 - 167971151 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001008.1 NP_001001008.1 1 - 1 481 126

View Full Product Details