The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012460.1 | NP_001012478.1 | 4 - 5 | 352 | 88 |
GenBank mRNA | BC089897.1 | - | 4 - 5 | 352 | 88 |