Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ckap4
Entrez Gene ID
362859
Gene Name
cytoskeleton-associated protein 4
Gene Aliases
-
Chromosome Location
Chr.7: 24935243 - 24947595 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108740.1 NP_001102210.1 1 - 2 174 79
XM_008765245.1 XP_008763467.1 1 - 2 807 79

View Full Product Details