Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mterfd3
Entrez Gene ID
366856
Gene Name
MTERF domain containing 3
Gene Aliases
RGD1311836, mTERF2
Chromosome Location
Chr.7: 24638208 - 24648263 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014265.2 NP_001014287.1 3 - 4 241 78
GenBank mRNA BC098693.1 - 3 - 4 241 78

View Full Product Details