Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nuak1
Entrez Gene ID
299694
Gene Name
NUAK family, SNF1-like kinase, 1
Gene Aliases
RGD1309956
Chromosome Location
Chr.7: 25039336 - 25111118 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106774.1 NP_001100244.1 3 - 4 675 110

View Full Product Details