Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Caps2
Entrez Gene ID
366891
Gene Name
calcyphosine 2
Gene Aliases
-
Chromosome Location
Chr.7: 54859088 - 54917406 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108990.1 NP_001102460.1 8 - 9 834 100
XM_006241337.2 XP_006241399.1 18 - 19 2182 100
XM_008765359.1 XP_008763581.1 17 - 18 1714 100

View Full Product Details