Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Folr2
Entrez Gene ID
293154
Gene Name
folate receptor 2 (fetal)
Gene Aliases
-
Chromosome Location
Chr.1: 166915045 - 166933377 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229859.2 XP_006229921.1 1 - 2 149 135

View Full Product Details