Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nrip3
Entrez Gene ID
361625
Gene Name
nuclear receptor interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.1: 174385424 - 174411141 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108498.1 NP_001101968.1 6 - 7 780 107

View Full Product Details