Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfeb
Entrez Gene ID
316214
Gene Name
transcription factor EB
Gene Aliases
Tcfeb
Chromosome Location
Chr.9: 15208141 - 15263966 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006244442.1 XP_006244504.1 1 - 2 143 54

View Full Product Details