The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014099.1 | NP_001014121.1 | 2 - 3 | 401 | 124 |
GenBank mRNA | BC079247.1 | - | 2 - 3 | 401 | 124 |