Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efhc1
Entrez Gene ID
301295
Gene Name
EF-hand domain (C-terminal) containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 27068505 - 27107713 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122947.2 NP_001116419.2 3 - 4 615 104

View Full Product Details