Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a27
Entrez Gene ID
85262
Gene Name
solute carrier family 25, member 27
Gene Aliases
Ucp4
Chromosome Location
Chr.9: 19880469 - 19907506 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053500.1 NP_445952.1 8 - 9 947 147
GenBank mRNA AJ300162.1 - 8 - 9 947 147

View Full Product Details