The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053603.2 | NP_446055.1 | 5 - 6 | 845 | 93 |
GenBank mRNA | AF323174.1 | - | 5 - 6 | 853 | 93 |