Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Api5
Entrez Gene ID
362170
Gene Name
apoptosis inhibitor 5
Gene Aliases
-
Chromosome Location
Chr.3: 83310878 - 83336053 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127379.1 NP_001120851.1 10 - 11 1290 141
XM_006234586.1 XP_006234648.1 10 - 11 1383 141
GenBank mRNA BC162003.1 - 10 - 11 1290 141

View Full Product Details