Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Astl
Entrez Gene ID
296129
Gene Name
astacin-like metalloendopeptidase (M12 family)
Gene Aliases
RGD1562279
Chromosome Location
Chr.3: 119783717 - 119799450 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106504.1 NP_001099974.1 5 - 6 510 98
XM_008762157.1 XP_008760379.1 5 - 6 453 98

View Full Product Details