Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fibin
Entrez Gene ID
499856
Gene Name
fin bud initiation factor homolog (zebrafish)
Gene Aliases
-
Chromosome Location
Chr.3: 101545207 - 101547478 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025042.2 NP_001020213.1 1 - 1 1998 97
GenBank mRNA BC085873.1 - 1 - 1 1942 97

View Full Product Details