The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013078.1 | NP_001013096.1 | 10 - 11 | 1400 | 66 |
XM_006234734.2 | XP_006234796.1 | 10 - 11 | 1438 | 66 | |
XM_006234735.2 | XP_006234797.1 | 10 - 11 | 1435 | 66 | |
XM_008762090.1 | XP_008760312.1 | 11 - 12 | 1381 | 66 | |
GenBank mRNA | BC082080.1 | - | 10 - 11 | 1400 | 66 |