Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxo6
Entrez Gene ID
192351
Gene Name
F-box protein 6
Gene Aliases
Fbg2, Fbxo6b
Chromosome Location
Chr.5: 164956313 - 164962072 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138917.3 NP_620272.3 4 - 5 641 70
XM_006239372.2 XP_006239434.1 5 - 6 945 70
XM_006239373.2 XP_006239435.1 5 - 6 878 70
XM_006239374.2 XP_006239436.1 5 - 6 805 70
XM_006239375.2 XP_006239437.1 5 - 6 809 70
GenBank mRNA AF393484.1 - 4 - 5 641 70
FQ232698.1 - 5 - 6 711 70

View Full Product Details