Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tprg1l
Entrez Gene ID
687090
Gene Name
tumor protein p63 regulated 1-like
Gene Aliases
mover
Chromosome Location
Chr.5: 171456299 - 171459488 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110318.2 NP_001103788.1 3 - 4 774 68
GenBank mRNA BC160865.1 - 3 - 4 623 68
EU220430.1 - 3 - 4 774 68

View Full Product Details