Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmgcl
Entrez Gene ID
79238
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase
Gene Aliases
-
Chromosome Location
Chr.5: 154294841 - 154308582 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024386.1 NP_077362.1 7 - 8 764 55
GenBank mRNA BC061797.1 - 7 - 8 766 55
FQ234382.1 - 7 - 8 762 55
Y10054.1 - 7 - 8 764 55

View Full Product Details