Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfap2e
Entrez Gene ID
313596
Gene Name
transcription factor AP-2 epsilon
Gene Aliases
RGD1563297, Tcfap2e
Chromosome Location
Chr.5: 144740950 - 144761787 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191764.1 NP_001178693.1 5 - 6 789 77

View Full Product Details