Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim63
Entrez Gene ID
140939
Gene Name
tripartite motif containing 63, E3 ubiquitin protein ligase
Gene Aliases
Murf, Murf1, Rnf28
Chromosome Location
Chr.5: 152533362 - 152547138 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080903.1 NP_543179.1 6 - 7 953 80
GenBank mRNA AY059627.1 - 6 - 7 953 80
BC061824.1 - 6 - 7 940 80

View Full Product Details