Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A3galt2
Entrez Gene ID
171553
Gene Name
alpha 1,3-galactosyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.5: 146933592 - 146945882 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138524.2 NP_612533.1 2 - 3 177 87
GenBank mRNA AF248543.1 - 2 - 3 176 87
BC061714.1 - 2 - 3 177 87

View Full Product Details