The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100872.1 | NP_001094342.1 | 7 - 8 | 1704 | 62 |
XM_006240303.2 | XP_006240365.1 | 7 - 8 | 1927 | 62 |