Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Erh
Entrez Gene ID
681415
Gene Name
enhancer of rudimentary homolog (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.6: 104280390 - 104290905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240290.2 XP_006240352.1 1 - 2 105 75

View Full Product Details