The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044290.2 | NP_001037755.2 | 10 - 11 | 1494 | 92 |
GenBank mRNA | BC083898.1 | - | 10 - 11 | 1431 | 92 |