Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpn2
Entrez Gene ID
362614
Gene Name
GPN-loop GTPase 2
Gene Aliases
Atpbd1b, RGD1311749
Chromosome Location
Chr.5: 151806458 - 151813954 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270959.1 NP_001257888.1 3 - 4 726 67

View Full Product Details